Learn More
Electron Microscopy Sciences Graphene on Thermal SiO2 on Ultra-Flat Silicon, 2 layers, 0.7 nm thick, 92.7% Transparency
SDP
Catalog No. 501929823
Encompass_Preferred
CVD graphene on ultra-flat thermal SiO2. The high quality graphene is produced by chemical vapor deposition, a process by which gaseous reactants are deposited onto a substrate. The Ultra-flat Thermal SiO2 Substrate consists of a 200 nm thermally grown SiO2 film on an ultra-flat silicon wafer with a normal thickness of 675 um. The size is 5 mm x 5 mm. Graphene coverage of the TEM grid is better than 75 percent. Available in four thicknesses of CVD graphene, 1, 2, 3-5, and 6-8.
Layers: 2 layers
Thickness: 0.7 nm
Transparency: 92.7%
Catalog No. 50-192-9823 Supplier Electron Microscopy Sciences Supplier No. 2GUFSIO25
CVD graphene on ultra-flat thermal SiO2. The high quality graphene is produced by chemical vapor deposition, a process by which gaseous reactants are deposited onto a substrate. The Ultra-flat Thermal SiO2 Substrate consists of a 200 nm thermally grown SiO2 film on an ultra-flat silicon wafer with a normal thickness of 675 um. The size is 5 mm x 5 mm. Graphene coverage of the TEM grid is better than 75 percent. Available in four thicknesses of CVD graphene, 1, 2, 3-5, and 6-8.
Layers: 2 layers
Thickness: 0.7 nm
Transparency: 92.7%