Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphic Controls Pen

Catalog No. 13939368
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Purple
Red
Quantity:
5 Pk.
6 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
13-939-368 Red 5 Pk.
13-939-287 Purple 6 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-939-368 Supplier Graphic Controls Supplier No. 10554047
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Type Graphic Controls Pen
Color Red
For Use With (Application) For chart recorders
Quantity 5 Pk.
Length (English) 0.6 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.