Learn More
Cole-Parmer AEMC 5070 5000 Volt Graphical Megaohmmeter with DataView Software
Catalog No. 50992625 Shop All Cole-Parmer Products
Encompass
This True Megohmmeterhas test voltage combinations of 500V, 1000V, 2500V and 5000Vand insulation measurements from 30KO to 10,000GO (10TO). It also has selectable and programmable test voltage (40 to 5100V); automatic calculation of DAR, PI and DD ratios; direct measurement and display of capacitance and leakage current, and display resistance, test voltage and run time. In addition, it has programmable test run times and PI ratio times; smooth and alarm functions;automatic test inhibition if live sample (>25V); automatic discharge and display of discharge voltage;large dual display with time, voltage and measurements shown (Models AE213020 and AE213021); graphic and digital display of test voltage, resistance and more (Model AE213030);bright blue electroluminescent backlight; programmable test voltage lock-out; programmable alarm setting, and auto power-down when not in use. The Megohmmeter is AC or DC powered with rechargeable NiMH batteries
Catalog No. 50-992-625 Supplier Cole-Parmer Supplier No. 2001829
This True Megohmmeterhas test voltage combinations of 500V, 1000V, 2500V and 5000Vand insulation measurements from 30KO to 10,000GO (10TO). It also has selectable and programmable test voltage (40 to 5100V); automatic calculation of DAR, PI and DD ratios; direct measurement and display of capacitance and leakage current, and display resistance, test voltage and run time. In addition, it has programmable test run times and PI ratio times; smooth and alarm functions;automatic test inhibition if live sample (>25V); automatic discharge and display of discharge voltage;large dual display with time, voltage and measurements shown (Models AE213020 and AE213021); graphic and digital display of test voltage, resistance and more (Model AE213030);bright blue electroluminescent backlight; programmable test voltage lock-out; programmable alarm setting, and auto power-down when not in use. The Megohmmeter is AC or DC powered with rechargeable NiMH batteries