Learn More
Glass Expansion Graphite Gasket (PKT 3) Shimadzu 2030/2040/2050 ICP-MS Sampler Cone (211-90174-41)
SDP
Catalog No. 502702941
Encompass_Preferred
Graphite Sampler Cone Gasket, PKT 3 (211-90174-41). Package of 3 graphite gaskets for Shimadzu 2030 ICP-MS sampler cone. Use a new gasket each time the cone is installed, even if it is the same cone, to ensure a proper seal. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When cones are not in use, keep them stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2941 Supplier Glass Expansion Supplier No. SZ5001
Graphite Sampler Cone Gasket, PKT 3 (211-90174-41). Package of 3 graphite gaskets for Shimadzu 2030 ICP-MS sampler cone. Use a new gasket each time the cone is installed, even if it is the same cone, to ensure a proper seal. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When cones are not in use, keep them stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001