Learn More
MSC Sprayon Dry Film Graphite Lubricant, Food Grade, 10 oz
Catalog No. 501112901
Encompass
Sprayon LU204 Dry Film Graphite Lubricant is an aerosol designed for high lubricity and extreme pressure applications. It quickly dries at room temperature, providing clean, long-wearing lubrication.
  • Medium pressure, high lubricity graphite
  • Adheres to most surfaces with minimal preparation
  • Low friction coefficient of 0.123
  • Dries rapidly at room temperature
  • Excellent conductivity
  • Provides clean, long-wearing lubrication
  • Food grade and NSF H2 registered
Catalog No. 50-111-2901 Supplier MSC Supplier No. 00252007
Sprayon LU204 Dry Film Graphite Lubricant is an aerosol designed for high lubricity and extreme pressure applications. It quickly dries at room temperature, providing clean, long-wearing lubrication.
  • Medium pressure, high lubricity graphite
  • Adheres to most surfaces with minimal preparation
  • Low friction coefficient of 0.123
  • Dries rapidly at room temperature
  • Excellent conductivity
  • Provides clean, long-wearing lubrication
  • Food grade and NSF H2 registered