Learn More
Perkin Elmer US LLC Graphite Capillary Column Ferrules - 0.125 in Nut x 1.0 mm I.D., Pkg. 10
Catalog No. 502113719
Encompass_Preferred
Recommended for high-temperature applications up to 450 °C. Not for GC/MS.
Catalog No. 50-211-3719 Supplier Perkin Elmer US LLC Supplier No. 09903394
Recommended for high-temperature applications up to 450 °C. Not for GC/MS.