Learn More
Grainger Graphite Conductivity Sensor ATC
Catalog No. 502767675
Encompass_Preferred
The SENSOREX CS150TC-K-1.0-TC-PT1 Graphite Thermocouple Probe is a 1.0 mm diameter K-type thermocouple probe with a graphite coating. It has a temperature range of -40 to 1250°C (-40 to 2282°F) and features a quick disconnect mini connector. The probe's graphite coating helps prevent corrosion and oxidation at high temperatures. With a response time of under 5 seconds, this durable probe is designed for continuous temperature measurement in kilns, furnaces, and other high temperature environments. The mini connector allows for easy replacement or detachment from thermocouple wires.
Catalog No. 50-276-7675 Supplier Grainger Supplier No. 16Z056
The SENSOREX CS150TC-K-1.0-TC-PT1 Graphite Thermocouple Probe is a 1.0 mm diameter K-type thermocouple probe with a graphite coating. It has a temperature range of -40 to 1250°C (-40 to 2282°F) and features a quick disconnect mini connector. The probe's graphite coating helps prevent corrosion and oxidation at high temperatures. With a response time of under 5 seconds, this durable probe is designed for continuous temperature measurement in kilns, furnaces, and other high temperature environments. The mini connector allows for easy replacement or detachment from thermocouple wires.