Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphite electrode, counter-flat top, 3.05mm dia, 38.10 mm long, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog No. AA14754KN
Change view
Click to view available options
Quantity:
100 Pc.
Packaging:
Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA14754KN 100 Pc. Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA14754KN Supplier Thermo Scientific Chemicals Supplier No. 014754KN
Add to Cart

missing translation for 'validMainframeMsg'

 

It is used as homogeneous catalysts. They are widely used in electric arc furnaces (EAF) and ladle furnaces (LF) for the melting of steel, non-ferrous metals and some non-metallic minerals. Graphite is used widely as an engineering material over a variety of applications. Applications include piston rings, thrust bearings, journal bearings and vanes. Carbon based seals are used in the shafts and fuel pumps of many aircraft jet engines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Graphite electrode
Packaging Tube
Quantity 100 Pc.
Solubility Information Insoluble in water.
Percent Purity 99.9995%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.