Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphite electrode, counter-spherical tip, 6.15mm dia, 38.10 mm long, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog No. AA14751KN
Change view
Click to view available options
Quantity:
100 Pc.
Packaging:
Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA14751KN 100 Pc. Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA14751KN Supplier Thermo Scientific Chemicals Supplier No. 014751KN
Add to Cart

 

The surface modification of graphite felt with concentrated sulfuric acid has led to dramatic improvement in the electroactivity of this material in the vanadium redox cell. A cell resistance of 2.50cm2 and energy efficiency of 91% were obtained in the vanadium cell, after the graphite felt electrode material was treated with boiling concentrated sulfuric acid for 5 hour. Graphite felt treated thermally at 400°C exhibited the greatest improvement in performance of the vanadium cell. Graphite electrode are used in primarily in electric furnace steel manufacturing. It can provide high level of electric conductivity and capability of sustaining the extremely high level of generating heat. It is also used in the refinement of steel and similar smelting process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Graphite electrode
Packaging Tube
Quantity 100 Pc.
Solubility Information Insoluble in water. Poorly soluble in general solvents. Soluble in molten nickel.
Percent Purity 99.9995%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.