Learn More
Perkin Elmer US LLC Graphite Ferrules - 0.0625 in Nut x 0.5 mm I.D.
Catalog No. 502112690
Encompass_Preferred
Recommended for high-temperature applications up to 450 °C. Not for GC/MS.
Catalog No. 50-211-2690 Supplier Perkin Elmer US LLC Supplier No. 04972433
Recommended for high-temperature applications up to 450 °C. Not for GC/MS.