Learn More
Perkin Elmer US LLC Vespel Ferrules - 1.6 mm I.D., Pkg. 5
Catalog No. 509052105
Encompass_Preferred
For use with 1/16 inch metal tubing and glass-lined recievers. Temperature limit is 350 °C.
Catalog No. 50-905-2105 Supplier Perkin Elmer US LLC Supplier No. L1003026
For use with 1/16 inch metal tubing and glass-lined recievers. Temperature limit is 350 °C.