Learn More
Glass Expansion Graphite Gasket for Agilent 7700/7900/8800/8900 Sampler Cone, PKT 3 (G3280-67009)
SDP
Catalog No. 501907580
Encompass_Preferred
Graphite Sampler Cone Gasket, PKT 3 (G3280-67009). For Agilent 7700/7900/8800/8900 ICP-MS. Use a new gasket each time the cone is installed, even if it is the same cone.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7580 Supplier Glass Expansion Supplier No. AT7703
Graphite Sampler Cone Gasket, PKT 3 (G3280-67009). For Agilent 7700/7900/8800/8900 ICP-MS. Use a new gasket each time the cone is installed, even if it is the same cone.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001