Learn More
TA Instruments Graphite Lids, DSC Graphite Lids, Materials That Might Alloy with Metals, Use with Non Oxidizing Purge Gas, Use with Graphite Sample Pans, 10 Pack
Catalog No. 5000580650 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Graphite Lids are DSC graphite lids used for evaluating materials that might alloy with metal pans. They must be used with a non-oxidizing purge gas such as nitrogen and are designed for use with graphite sample pans (P/N: 900874.901).
Catalog No. 50-005-80650 Supplier TA Instruments Supplier No. 900873.901
Waters TA Instruments Graphite Lids are DSC graphite lids used for evaluating materials that might alloy with metal pans. They must be used with a non-oxidizing purge gas such as nitrogen and are designed for use with graphite sample pans (P/N: 900874.901).