Learn More
Perkin Elmer US LLC Graphite One-Piece Ferrules - 0.25 in Nut x 0.25 in I.D., Pkg. 10
Supplier:  Perkin Elmer US LLC 09920140
Encompass_Preferred
Recommended for high-temperature applications up to 450 °C. Not for GC/MS.
Catalog No. 50-211-7112