Learn More
Perkin Elmer US LLC Graphite One-Piece Ferrules - 0.0625 in Nut x 0.8 mm I.D., Pkg. 10
Catalog No. 502113091
Encompass_Preferred
Recommended for high-temperature applications up to 450 °C. Not for GC/MS.
Catalog No. 50-211-3091 Supplier Perkin Elmer US LLC Supplier No. 09920141
Recommended for high-temperature applications up to 450 °C. Not for GC/MS.