Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphite plate, highly oriented pyrolytic graphite (HOPG), 10x10x1mm (0.394x0.394x0.039in), Thermo Scientific Chemicals

Catalog No. AA43836KF
Change view
Click to view available options
Quantity:
1 Pc.
Packaging:
Shrink-wrapped
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA43836KF 1 Pc. Shrink-wrapped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA43836KF Supplier Thermo Scientific Chemicals Supplier No. 043836KF
Add to Cart

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Graphite plate
Packaging Shrink-wrapped
Quantity 1 Pc.
MDL Number MFCD00144065

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.