Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphite powder, microcrystalline, -325 mesh, 75-82% C, 18-25% Ash, Thermo Scientific Chemicals

Catalog No. p-7048311
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1013014 25 g
AA1013030 250 g
AA10130A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1013014 Supplier Thermo Scientific Chemicals Supplier No. 01013014
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Mesh Size −325 Mesh
Odor Odorless
Quantity 25 g
UN Number UN1325
Physical Form Powder
Chemical Name or Material Graphite powder
Hazard Category H228-H319-H335
Hazard Statement GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 231-955-3
RTECSNumber MD9659600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.