Learn More
Electron Microscopy Sciences Carbon (Graphite) Rods Spectro-Grade, ⅛" diameter, 12" OAL
SDP
Catalog No. 5024904
Encompass_Preferred
Carbon Rods (graphite) with an impurity level of 2ppm or less with a maximum level element of each of 1 ppm. Ideal for all TEM, SEM and other critical applications.
Diameter: 1/8"
OAL: 12"
Catalog No. 50-249-04 Supplier Electron Microscopy Sciences Supplier No. 70200
Carbon Rods (graphite) with an impurity level of 2ppm or less with a maximum level element of each of 1 ppm. Ideal for all TEM, SEM and other critical applications.
Diameter: 1/8"
OAL: 12"