Learn More
TA Instruments Graphite Sample Pans, DSC Graphite Pans, Materials That Might Alloy with Metals, Use with Non Oxidizing Purge Gas, Use with Graphite Lids, 10 Pack
Catalog No. 5000580813 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Graphite Sample Pans are DSC graphite pans used for evaluating materials that might alloy with metal pans. They must be used with a non-oxidizing purge gas such as nitrogen and are designed for use with graphite lids (P/N: 900873.901).
Catalog No. 50-005-80813 Supplier TA Instruments Supplier No. 900874.901
Waters TA Instruments Graphite Sample Pans are DSC graphite pans used for evaluating materials that might alloy with metal pans. They must be used with a non-oxidizing purge gas such as nitrogen and are designed for use with graphite lids (P/N: 900873.901).