Learn More
Electron Microscopy Sciences Graphite Spray Adhesive 450 GR
SDP
Catalog No. 50980319
Encompass_Preferred
An easy to use graphite spray to coat small samples. It is fast drying and produces a very flat, thin, and uniform graphite film. Its service temperature is up to 204 degree C.  It's electrical resistance is 1.2 kohm/sq" at 1 ml thickness.
Catalog No. 50-980-319 Supplier Electron Microscopy Sciences Supplier No. 12648
An easy to use graphite spray to coat small samples. It is fast drying and produces a very flat, thin, and uniform graphite film. Its service temperature is up to 204 degree C.  It's electrical resistance is 1.2 kohm/sq" at 1 ml thickness.