Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphite tape, square weave, 0.56mm (0.022 in.) thick x 100mm (3.94in) wide, 99.9%, Thermo Scientific™

Catalog No. AA10136BY
Change view
Click to view available options
Quantity:
25 cm
100 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10136BY 100 cm
AA10136BU 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA10136BY Supplier Thermo Scientific Chemicals Supplier No. 10136BY
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00144065

Specifications

Quantity 100 cm
Percent Purity 99.9%
Chemical Name or Material Graphite tape, Square weave, 0.56mm (0.022 in.) thick x 100mm (3.94 in.) wide
EINECSNumber 231-955-3
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.