Learn More
Thermo Scientific Seal, Graphite/Vespel
Supplier:  Thermo Scientific 7000520922
Encompass
Graphite and vespel seal, provides chemical resistance, low friction, and high temperature capabilities as a replacement analytical instrument seal to maintain vacuum by creating a tight junction between surfaces.
Catalog No. NC1506017