Learn More
Perkin Elmer US LLC Graphite/Vespel Capillary Column Ferrules - 0.0625 in Nut x 0.8 mm I.D., Pkg. 10
Catalog No. 502117122
Encompass_Preferred
Recommended for use with injectors and detectors at atmospheric pressure and at temperatures up to 350 °C.
Catalog No. 50-211-7122 Supplier Perkin Elmer US LLC Supplier No. 09920107
Recommended for use with injectors and detectors at atmospheric pressure and at temperatures up to 350 °C.