Learn More
Perkin Elmer US LLC Graphite/Vespel Capillary Column Ferrules - 0.0625 in Nut x 0.4 mm I.D., Pkg. 10
Catalog No. 502117115
Encompass_Preferred
Recommended for use with injectors and detectors at atmospheric pressure and at temperatures up to 350 °C.
Catalog No. 50-211-7115 Supplier Perkin Elmer US LLC Supplier No. 09920104
Recommended for use with injectors and detectors at atmospheric pressure and at temperatures up to 350 °C.