Learn More
Perkin Elmer US LLC Graphite/Vespel Ferrules - 0.125 in Nut x 0.125 in I.D., Pkg. 10
Catalog No. 502112429
Encompass_Preferred
Recommended for use with Detectors/Injectors at temperatures up to 350 °C.
Catalog No. 50-211-2429 Supplier Perkin Elmer US LLC Supplier No. N9302081
Recommended for use with Detectors/Injectors at temperatures up to 350 °C.