Learn More
Perkin Elmer US LLC Graphite/Vespel Ferrules - 0.25 in Nut x 0.25 in I.D., Pkg. 10
Catalog No. 502113731
Encompass_Preferred
Recommended for use with Detectors/Injectors at temperatures up to 350 °C.
Catalog No. 50-211-3731 Supplier Perkin Elmer US LLC Supplier No. 09903739
Recommended for use with Detectors/Injectors at temperatures up to 350 °C.