Learn More
Medchemexpress LLC Graphitic carbon nitride | 143334-20-7 | MFCD00040542 | 97.0% | (C3H4)n | 500 MG
SDP
Catalog No. 5000284186
Encompass_Preferred
Graphitic carbon nitride is a metal-free, polymeric photocatalyst (g-C3N4) supplied as a light yellow solid. It exhibits semiconductor optical absorption near 460 nm and is intended for materials and photocatalysis research applications. The material is provided for research use only and is accompanied by storage guidance for solid and solvent conditions.
  • Metal-free photocatalyst suitable for visible-light driven reactions.
  • Semiconductor absorption near 460 nm.
  • Polymeric composition expressed as (C3H4)n.
  • High purity (97%).
  • Solid powder; light yellow to yellow in color.
  • Stable at room temperature for up to 3 years; in solvent, store at -80°C for 2 years or -20°C for 1 year.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-84186 Supplier Medchemexpress LLC Supplier No. HYW540000500MG
Graphitic carbon nitride is a metal-free, polymeric photocatalyst (g-C3N4) supplied as a light yellow solid. It exhibits semiconductor optical absorption near 460 nm and is intended for materials and photocatalysis research applications. The material is provided for research use only and is accompanied by storage guidance for solid and solvent conditions.
  • Metal-free photocatalyst suitable for visible-light driven reactions.
  • Semiconductor absorption near 460 nm.
  • Polymeric composition expressed as (C3H4)n.
  • High purity (97%).
  • Solid powder; light yellow to yellow in color.
  • Stable at room temperature for up to 3 years; in solvent, store at -80°C for 2 years or -20°C for 1 year.
  • For research use only; not for human or clinical use.