Learn More
Electron Microscopy Sciences Graphitized Carbon Black Crystals
SDP
Catalog No. 5026886
Encompass_Preferred
Since crystal lattice plane spacings are accurately known from x-ray measurements, they provide a good test of microscope stability and a calibration of magnification in the upper range of magnification of the instrument. Plane Spacing 0.34
Catalog No. 50-268-86 Supplier Electron Microscopy Sciences Supplier No. 80037
Since crystal lattice plane spacings are accurately known from x-ray measurements, they provide a good test of microscope stability and a calibration of magnification in the upper range of magnification of the instrument. Plane Spacing 0.34