Learn More
Hampton Research GRAS Screen 5, 1 mL, Deep Well block format
SDP
Catalog No. NC1527509
Encompass
GRAS Screen 5 and 6 were developed by Hampton Research for the crystallization of proteins, including monoclonal antibodies.
Catalog No. NC1527509 Supplier Hampton Research Supplier No. HR2455
GRAS Screen 5 and 6 were developed by Hampton Research for the crystallization of proteins, including monoclonal antibodies.