Learn More
Med Vet International Humaniac Cat Grasper 28", J0141AN, Each
Catalog No. 5000495948
Encompass_Preferred
This lightweight grasper was designed for effective, but safe, control of cats. It is ideal for reaching into the back of a cage to secure a fractious feline. The smooth covered jaws are plastisol covered to minimize discomfort. If necessary, the animal can be quickly released. Shaft is 28" long.
Catalog No. 50-004-95948 Supplier Med Vet International Supplier No. JOR141AN
This lightweight grasper was designed for effective, but safe, control of cats. It is ideal for reaching into the back of a cage to secure a fractious feline. The smooth covered jaws are plastisol covered to minimize discomfort. If necessary, the animal can be quickly released. Shaft is 28" long.