Learn More
Electron Microscopy Sciences Integrated EyePiece (Simplified) Graticle G50, 23mm
SDP
Catalog No. 501902061
Encompass_Preferred
The eyepiece reticle has a 100 point integrating pattern. It is a slightly simplified version of G47 pattern.
Size: 23 mm
Catalog No. 50-190-2061 Supplier Electron Microscopy Sciences Supplier No. 6800523
The eyepiece reticle has a 100 point integrating pattern. It is a slightly simplified version of G47 pattern.
Size: 23 mm