Learn More
Electron Microscopy Sciences Pharmaceutical PSA Pattern Graticle G-57, 23 mm, Glass sandwich
SDP
Catalog No. 501928029
Encompass_Preferred
This graticule was designed for the pharmaceutical industry. However, it is also useful where particle size considerations are restricted to 10 mm and 25 mm. Dots and circles give quick references for these two sizes. In addition a scale is incorporated. The microscope must be calibrated when ordering this graticule, such that the circle must equate to 1 mm on the microscope stage.
Size: 23 mm
Surface: Glass sandwich
Catalog No. 50-192-8029 Supplier Electron Microscopy Sciences Supplier No. 6809123
This graticule was designed for the pharmaceutical industry. However, it is also useful where particle size considerations are restricted to 10 mm and 25 mm. Dots and circles give quick references for these two sizes. In addition a scale is incorporated. The microscope must be calibrated when ordering this graticule, such that the circle must equate to 1 mm on the microscope stage.
Size: 23 mm
Surface: Glass sandwich