Learn More
Electron Microscopy Sciences Microscope Graticule NE1 Horizontal, 24.5 mm, 10 mm with 0.1 mm subdivisions
SDP
Catalog No. 501927983
Encompass_Preferred
These scales are used for the measuring of lengths of specimen or distances between points on a variety of different shaped objects. By dividing the division of the chosen graticule by the magnification of the objective one obtains an approximate value that each division will represent on the stage. Vertical micrometer scale.
Size: 24.5 mm
Catalog No. 50-192-7983 Supplier Electron Microscopy Sciences Supplier No. 6801024.5
These scales are used for the measuring of lengths of specimen or distances between points on a variety of different shaped objects. By dividing the division of the chosen graticule by the magnification of the objective one obtains an approximate value that each division will represent on the stage. Vertical micrometer scale.
Size: 24.5 mm