Learn More
Electron Microscopy Sciences Indexed Grid Graticle NE10A, 16 mm, 5 x 5 mm with 0.5 mm pitch
SDP
Catalog No. 5028860
Encompass_Preferred
Useful for particle counting, particularly when reference is needed between workers. Also, they are useful for area of specimen determinations. Indexed grid marked 1-10 and A-J.
Size: 16 mm
Divisions: 5 x 5 mm with 0.5 mm pitch
Surface: Chrome image
Catalog No. 50-288-60 Supplier Electron Microscopy Sciences Supplier No. 6801116A
Useful for particle counting, particularly when reference is needed between workers. Also, they are useful for area of specimen determinations. Indexed grid marked 1-10 and A-J.
Size: 16 mm
Divisions: 5 x 5 mm with 0.5 mm pitch
Surface: Chrome image