Learn More
Electron Microscopy Sciences Indexed Grid Graticle NE11A, 16 mm, 10 x 10 mm with 1.0 mm pitch
SDP
Catalog No. 5028866
Encompass_Preferred
Useful for particle counting, particularly when reference is needed between workers. Also, they are useful for area of specimen determinations. Indexed grid marked 1-10 and A-J.
Size: 16 mm
Divisions: 10 x 10 mm with 1.0 mm pitch
Surface: Chrome image
Catalog No. 50-288-66 Supplier Electron Microscopy Sciences Supplier No. 6801616
Useful for particle counting, particularly when reference is needed between workers. Also, they are useful for area of specimen determinations. Indexed grid marked 1-10 and A-J.
Size: 16 mm
Divisions: 10 x 10 mm with 1.0 mm pitch
Surface: Chrome image