Learn More
Electron Microscopy Sciences Indexed Grid Graticle NE11A, 26 mm, 10 x 10 mm with 1.0 mm pitch
SDP
Catalog No. 501928002
Encompass_Preferred
Useful for particle counting, particularly when reference is needed between workers. Also, they are useful for area of specimen determinations. Indexed grid marked 1-10 and A-J.
Size: 26 mm
Divisions: 10 x 10 mm with 1.0 mm pitch
Surface: Chrome image
Catalog No. 50-192-8002 Supplier Electron Microscopy Sciences Supplier No. 6801626
Useful for particle counting, particularly when reference is needed between workers. Also, they are useful for area of specimen determinations. Indexed grid marked 1-10 and A-J.
Size: 26 mm
Divisions: 10 x 10 mm with 1.0 mm pitch
Surface: Chrome image