Learn More
Electron Microscopy Sciences Microscope Graticule NE120 Horizontal, Custom, 20 mm with 100 divisions of 0.1 mm
SDP
Catalog No. 501901770
Encompass_Preferred
These scales are used for the measuring of lengths of specimen or distances between points on a variety of different shaped objects. By dividing the division of the chosen graticule by the magnification of the objective one obtains an approximate value that each division will represent on the stage. Horizontal micrometer scale.
Size: Custom
Catalog No. 50-190-1770 Supplier Electron Microscopy Sciences Supplier No. 6797650
These scales are used for the measuring of lengths of specimen or distances between points on a variety of different shaped objects. By dividing the division of the chosen graticule by the magnification of the objective one obtains an approximate value that each division will represent on the stage. Horizontal micrometer scale.
Size: Custom