Learn More
Electron Microscopy Sciences Crossed Scale Microscope Graticle NE17, 27 mm, 10 mm with 0.1 mm subdivisions
SDP
Catalog No. 501927991
Encompass_Preferred
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Crossed micrometer scale.
Size: 27 mm
Divisions: 10 mm with 0.1 mm subdivisions
Surface: Chrome image
Catalog No. 50-192-7991 Supplier Electron Microscopy Sciences Supplier No. 6798927
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Crossed micrometer scale.
Size: 27 mm
Divisions: 10 mm with 0.1 mm subdivisions
Surface: Chrome image