Learn More
Electron Microscopy Sciences Crossed Scale Microscope Graticle NE18, 16 mm, 5 mm with 0.05 mm subdivisions
SDP
Catalog No. 5025157
Encompass_Preferred
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis.
Size: 16 mm
Divisions: 5 mm with 0.05 mm subdivisions
Surface: Chrome image
Catalog No. 50-251-57 Supplier Electron Microscopy Sciences Supplier No. 6799416
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis.
Size: 16 mm
Divisions: 5 mm with 0.05 mm subdivisions
Surface: Chrome image