Learn More
Electron Microscopy Sciences Microscope Graticule NE20 Horizontal, 16 mm, 0.1 inch with 100 subdivisions
SDP
Catalog No. 5026571
Encompass_Preferred
These scales are used for the measuring of lengths of specimen or distances between points on a variety of different shaped objects. By dividing the division of the chosen graticule by the magnification of the objective one obtains an approximate value that each division will represent on the stage. Horizontal micrometer scale.
Size: 16 mm
Catalog No. 50-265-71 Supplier Electron Microscopy Sciences Supplier No. 6798716
These scales are used for the measuring of lengths of specimen or distances between points on a variety of different shaped objects. By dividing the division of the chosen graticule by the magnification of the objective one obtains an approximate value that each division will represent on the stage. Horizontal micrometer scale.
Size: 16 mm