Learn More
Electron Microscopy Sciences Microscope Graticule NE28 Horizontal, 21 mm, 1 mm with 100 subdivisions
SDP
Catalog No. 5026579
Encompass_Preferred
These scales are used for the measuring of lengths of specimen or distances between points on a variety of different shaped objects. By dividing the division of the chosen graticule by the magnification of the objective one obtains an approximate value that each division will represent on the stage. Vertical micrometer scale.
Size: 21 mm
Catalog No. 50-265-79 Supplier Electron Microscopy Sciences Supplier No. 6799221
These scales are used for the measuring of lengths of specimen or distances between points on a variety of different shaped objects. By dividing the division of the chosen graticule by the magnification of the objective one obtains an approximate value that each division will represent on the stage. Vertical micrometer scale.
Size: 21 mm