Learn More
Electron Microscopy Sciences Whipple Grid Graticle NE29, 24.5 mm, 100 squares in 7 mm
SDP
Catalog No. 501928012
Encompass_Preferred
Originally designed for water particle analysis, but may be used for other aspects of particle counting. Grid showed: Ratio of full square to smallest is 50:1. Area is 2500:1. Reference: Microscopy of Drinking Water.
Size: 24.5 mm
Divisions: 100 squares in 7 mm
Catalog No. 50-192-8012 Supplier Electron Microscopy Sciences Supplier No. 6800924.5
Originally designed for water particle analysis, but may be used for other aspects of particle counting. Grid showed: Ratio of full square to smallest is 50:1. Area is 2500:1. Reference: Microscopy of Drinking Water.
Size: 24.5 mm
Divisions: 100 squares in 7 mm