Learn More
Electron Microscopy Sciences Matthews Spray Droplet Graticle NE30, 23mm
SDP
Catalog No. 501901802
Encompass_Preferred
For size and distribution assessments of aersol droplets. Used in conjunction with a 40X Microscope for direct measurements of droplets from 50-400 micron diameters. The actual pattern sizes are 50, 100, 200, and 400 microns. W.H.O. (Details on Request) and G.A. Matthews. Imperial College.
Size: 23 mm
Catalog No. 50-190-1802 Supplier Electron Microscopy Sciences Supplier No. 6800323
For size and distribution assessments of aersol droplets. Used in conjunction with a 40X Microscope for direct measurements of droplets from 50-400 micron diameters. The actual pattern sizes are 50, 100, 200, and 400 microns. W.H.O. (Details on Request) and G.A. Matthews. Imperial College.
Size: 23 mm