Learn More
Electron Microscopy Sciences Concentric Circles Graticle NE48, 19mm, 10 circles 1 mm-10 mm
SDP
Catalog No. 501901795
Encompass_Preferred
Concentric circles eyepiece graticle with 10 circles 1 mm-10 mm, with graduated cross hairs.
Size: 19mm
Divisions: 10 circles 1 mm-10 mm
Catalog No. 50-190-1795 Supplier Electron Microscopy Sciences Supplier No. 6799119
Concentric circles eyepiece graticle with 10 circles 1 mm-10 mm, with graduated cross hairs.
Size: 19mm
Divisions: 10 circles 1 mm-10 mm