Learn More
Electron Microscopy Sciences Crossed Gauge Lines Microscope Graticule NE53, 16mm
SDP
Catalog No. 5025161
Encompass_Preferred
Use as crossed lines, but for the measuring of distances between thelines. Greater accuracy can be obtained by locatng the specimen detail between the graticules gauge pair.
Size: 16mm
Divisions: Two vertical lines 0.1 mm apart
Catalog No. 50-251-61 Supplier Electron Microscopy Sciences Supplier No. 6798616
Use as crossed lines, but for the measuring of distances between thelines. Greater accuracy can be obtained by locatng the specimen detail between the graticules gauge pair.
Size: 16mm
Divisions: Two vertical lines 0.1 mm apart