Learn More
Electron Microscopy Sciences Crossed Gauge Lines Microscope Graticule NE53, 19mm
SDP
Catalog No. 5025162
Encompass_Preferred
Use as crossed lines, but for the measuring of distances between thelines. Greater accuracy can be obtained by locatng the specimen detail between the graticules gauge pair.
Size: 19mm
Divisions: Two vertical lines 0.1 mm apart
Catalog No. 50-251-62 Supplier Electron Microscopy Sciences Supplier No. 6798619
Use as crossed lines, but for the measuring of distances between thelines. Greater accuracy can be obtained by locatng the specimen detail between the graticules gauge pair.
Size: 19mm
Divisions: Two vertical lines 0.1 mm apart