Learn More
Electron Microscopy Sciences Crossed Gauge Lines Microscope Graticule NE 54, 19mm
SDP
Catalog No. 501901765
Encompass_Preferred
Use as crossed lines, but for the measuring of distances between thelines. Greater accuracy can be obtained by locatng the specimen detail between the graticules gauge pair.
Size: 19mm
Divisions: Two vertical lines 0.2 mm apart
Catalog No. 50-190-1765 Supplier Electron Microscopy Sciences Supplier No. 6797519
Use as crossed lines, but for the measuring of distances between thelines. Greater accuracy can be obtained by locatng the specimen detail between the graticules gauge pair.
Size: 19mm
Divisions: Two vertical lines 0.2 mm apart