Learn More
Electron Microscopy Sciences Scale with Cross Line Microscope Graticle NE7, 24.5 mm, 10 mm with 0.1 mm spacing
SDP
Catalog No. 501927994
Encompass_Preferred
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Horizontal micrometer scale 10mm long with crosslines at 0.1mm spacing. Surface chrome image.
Size: 24.5 mm
Divisions: 10 mm with 0.1 mm spacing
Surface: Chrome image
Catalog No. 50-192-7994 Supplier Electron Microscopy Sciences Supplier No. 6801324.5
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Horizontal micrometer scale 10mm long with crosslines at 0.1mm spacing. Surface chrome image.
Size: 24.5 mm
Divisions: 10 mm with 0.1 mm spacing
Surface: Chrome image