Learn More
Electron Microscopy Sciences Crossed Scale Microscope Graticle NE70 (imperial), 23 mm, 0.8' with 400 divisions of 0.002"
SDP
Catalog No. 501901781
Encompass_Preferred
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Crossed Micrometer scale (imperial).
Size: 23 mm
Divisions: 0.8' with 400 divisions of 0.002"
Surface: Chrome image
Catalog No. 50-190-1781 Supplier Electron Microscopy Sciences Supplier No. 6797923
Used as a horizontal and a vertical scale, they are especially useful when interested in measurement in different axis. Crossed Micrometer scale (imperial).
Size: 23 mm
Divisions: 0.8' with 400 divisions of 0.002"
Surface: Chrome image