Learn More
Electron Microscopy Sciences Indexed Grid Graticle NE71, 23 mm, 20 x 20 Grid of 0.5 mm squares
SDP
Catalog No. 501901790
Encompass_Preferred
Useful for particle counting, particularly when reference is needed between workers. Also, they are useful for area of specimen determinations. Index Pattern.
Size: 23 mm
Divisions: 20 x 20 Grid of 0.5 mm squares
Catalog No. 50-190-1790 Supplier Electron Microscopy Sciences Supplier No. 6798123
Useful for particle counting, particularly when reference is needed between workers. Also, they are useful for area of specimen determinations. Index Pattern.
Size: 23 mm
Divisions: 20 x 20 Grid of 0.5 mm squares